Learn More
Abcam Plasmin Activity Assay Kit (Fluorometric)
Product Code. 32112047
Change view
Click to view available options
1x96 Tests
Product Code.
32112047 1x96 Tests
1 options
Product Code. 32112047 Supplier Abcam Supplier No. ab3246301x96Tests
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure Plasminogen activity in 1h in Biofluids with Plasmin Activity Assay Kit AB324630. Sensitivity = 8.66 μU. Use with Microplate (12 x 8 well strips).
Measure Plasminogen activity in 1h in Biofluids with Plasmin Activity Assay Kit AB324630. Sensitivity = 8.66 μU. Use with Microplate (12 x 8 well strips).
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1 x 96 Tests