Learn More
Medline Scientific™ Plan Objective
For use with Varizoom binocular microscopes.
Brand: Medline Scientific™ 7700.0020M
Additional Details : Weight : 1.00000kg
Description
- 0.5x increases working distance and increases field of view
- 2.0 x magnification
Specifications
Plan Objective | |
Varizoom Stereo Microscopes | |
2X | |
1 Ea. |