Learn More
Medline Scientific™ Plan Objective
For use with Varizoom binocular microscopes.
£1020.00 - £2555.00
Product Code | Brand | Magnification Power | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Magnification Power | Price | Quantity & Availability | |||||
12613035
|
Medline Scientific™
7700.0005 |
0.5X |
£1020.00
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
12835101
|
Medline Scientific™
7700.0020M |
2X |
£2555.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Description
- 0.5x increases working distance and increases field of view
- 2.0 x magnification
Specifications
Plan Objective | |
1 Ea. |