missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKLR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £255.00
Specifications
| Antigen | PKLR |
|---|---|
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:1000-1:4000, Western Blot (Negative) |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611382
|
Novus Biologicals
NBP2-97071-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608654
|
Novus Biologicals
NBP2-97071-50ul |
50 μg |
£178.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKLR Polyclonal antibody specifically detects PKLR in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot (Negative)Specifications
| PKLR | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Chaperone Mediated Autophagy (CMA), Lipid and Metabolism, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 5313 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:1000-1:4000, Western Blot (Negative) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.1.40, PK1PKRL, PKL, PKR, Pyruvate kinase 1, pyruvate kinase isozyme R/L, pyruvate kinase isozymes R/L, pyruvate kinase type L, pyruvate kinase, liver and blood cell, pyruvate kinase, liver and RBC, Red cell/liver pyruvate kinase, RPK, R-type/L-type pyruvate kinase | |
| Produced in rabbits immunized with purified, recombinant Human PKLR (Accession#: NP_000289.1; Leu47-Ser574) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title