missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKLR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97071-50ul
This item is not returnable.
View return policy
Description
PKLR Polyclonal antibody specifically detects PKLR in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot (Negative)
Specifications
| PKLR | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:1000-1:4000, Western Blot (Negative) | |
| EC 2.7.1.40, PK1PKRL, PKL, PKR, Pyruvate kinase 1, pyruvate kinase isozyme R/L, pyruvate kinase isozymes R/L, pyruvate kinase type L, pyruvate kinase, liver and blood cell, pyruvate kinase, liver and RBC, Red cell/liver pyruvate kinase, RPK, R-type/L-type pyruvate kinase | |
| Produced in rabbits immunized with purified, recombinant Human PKLR (Accession#: NP_000289.1; Leu47-Ser574) | |
| 50 μg | |
| Chaperone Mediated Autophagy (CMA), Lipid and Metabolism, Signal Transduction | |
| 5313 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction