Learn More
PKC Epsilon Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-20877-20UL
 View more versions of this product
Product Code. 18069365
  • £135.00 / 20µL
Description
The PKC Epsilon antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PKC Epsilon. This antibody recognizes human, mouse, rat antigen. The PKC Epsilon antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
PKC Epsilon
500 μg/mL
PKCE, PRKCE, Protein kinase C epsilon type, protein kinase C, epsilon
20 μL
5581
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa