Learn More
PKC Epsilon Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | PKC Epsilon |
|---|---|
| Concentration | 500 μg/mL |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18069365
|
Proteintech
CL488-20877-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18040336
|
Proteintech
CL488-20877-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PKC Epsilon antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PKC Epsilon. This antibody recognizes human, mouse, rat antigen. The PKC Epsilon antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| PKC Epsilon | |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse, Rat | |
| PKCE, PRKCE, Protein kinase C epsilon type, protein kinase C, epsilon | |
| Primary |
| 500 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 5581 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.