Learn More
Eppendorf™ Piston Spring For Research 3 Neo Single Channel Pipettes
Product Code. 30689738 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
10 mL
100 μL
2 mL
2.5 to 20 μL
200 μL
300 μL
5 mL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
30689738 200 μL Each
30689639 2 mL Each
30689734 5 mL Each
30689660 10 mL Each
30689673 2.5 to 20 μL Each
30689675 100 μL Each
30689608 300 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 30689738 Supplier Eppendorf™ Supplier No. 3102636005
Only null left
Add to basket
Add to basket
Piston springs provide resistance for controlled piston movement with consistent tactile feedback
  • Piston springs provide resistance for controlled piston movement, return the piston to its original position after dispensing
  • Ensure precision by offering consistent tactile feedback
  • Available in various Volume Capacity and color code
TRUSTED_SUSTAINABILITY
Specifications
For Use With Pipettes
Material Stainless Steel
Type Piston
Volume (Metric) 200 μL
For Use With (Equipment) Pipette Tips
Quantity 1 Pk.