Description
- Piston springs provide resistance for controlled piston movement, return the piston to its original position after dispensing
- Ensure precision by offering consistent tactile feedback
- Available in various Volume Capacity and color code
Specifications
Specifications
| For Use With | Pipettes |
| Material | Stainless Steel |
| Type | Piston |
| Volume (Metric) | 100 μL |
| For Use With (Equipment) | Pipette Tips |
| Quantity | 1 Pk. |