Learn More
Eppendorf™ Pipette Holder 2 for one Multipette™ E3/E3x or Multipette™ stream/Xstream Pipette
GREENER_CHOICE
Product Code. 15789488 Shop All Eppendorf Products
Click to view available options
:
Multipette™ E3/E3x or Multipette™ stream/Xstream Pipette
Product Code. 15789488
Brand: Eppendorf™ 3116000139
Only null left
Add to basket
Add to basket
Holder designed for wall mounting or for installation in Pipette Carousel 2
  • Carries one Multipette™ E3/E3x or Multipette™ stream/Xstream
  • Same as Charger Shell 2 for Multipette E3/E3x or Multipette stream/Xstream but without charging functionality
  • Designed for wall mounting or for installation in Pipette Carousel 2
  • Sticky tape for wall mounting included
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For one Multipette™ E3/E3x or Multipette™ stream/Xstream Pipette