missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PINCH1/LIMS1 Antibody (5G7), Novus Biologicals™
Mouse Monoclonal Antibody
£188.00 - £438.00
Specifications
| Antigen | PINCH1/LIMS1 |
|---|---|
| Clone | 5G7 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614456
|
Novus Biologicals
NBP2-37581-0.025ml |
0.025 mL |
£188.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18708943
|
Novus Biologicals
NBP2-37581 |
0.1 mL |
£438.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PINCH1/LIMS1 Monoclonal specifically detects PINCH1/LIMS1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| PINCH1/LIMS1 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with 0.03% Sodium Azide | |
| 3987 | |
| Purified recombinant fragment of human PINCH expressed in E. Coli. | |
| Primary | |
| 37 kDa |
| 5G7 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| LIM and senescent cell antigen-like domains 1, LIM and senescent cell antigen-like-containing domain protein 1, PINCH-1, PINCHPINCH1Particularly interesting new Cys-His protein 1, Renal carcinoma antigen NY-REN-48 | |
| LIMS1 | |
| IgG1 | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title