missing translation for 'onlineSavingsMsg'
Learn More

PINCH1/LIMS1 Antibody (5G7), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-37581-0.025ml

 View more versions of this product

Product Code. 18614456

  • £188.00 / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PINCH1/LIMS1 Monoclonal specifically detects PINCH1/LIMS1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PINCH1/LIMS1
Monoclonal
Unconjugated
Ascites with 0.03% Sodium Azide
LIMS1
Purified recombinant fragment of human PINCH expressed in E. Coli.
Unpurified
RUO
3987
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
IgG1
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
5G7
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000
LIM and senescent cell antigen-like domains 1, LIM and senescent cell antigen-like-containing domain protein 1, PINCH-1, PINCHPINCH1Particularly interesting new Cys-His protein 1, Renal carcinoma antigen NY-REN-48
Mouse
37 kDa
0.025 mL
Primary
Human
Ascites
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only