missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PILR-beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £470.00
Specifications
| Antigen | PILR-beta |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18458350
|
Novus Biologicals
NBP1-84701-25ul |
25 μL |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18244187
|
Novus Biologicals
NBP1-84701 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PILR-beta Polyclonal specifically detects PILR-beta in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PILR-beta | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 29990 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AGHPEIGEAAVAVHQGDQTHHHPGCHNHHHLEAQQHNHHSRPQGHRKQRALRIMAPKSGHCHQGCIGCRCAQNCHFGTAVPPPPVVEEKER | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| activating receptor PILRbeta, Activating receptor PILR-beta, Cell surface receptor FDFACT, cell surface receptor FDFACT1, cell surface receptor FDFACT2, FDFACT, FDFACT1, FDFACT2, paired immunoglobin-like receptor beta, paired immunoglobin-like type 2 receptor beta, paired immunoglobulin-like receptor beta, paired immunoglobulin-like type 2 receptor beta | |
| PILRB | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title