missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PILR-beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-84701
This item is not returnable.
View return policy
Description
PILR-beta Polyclonal specifically detects PILR-beta in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PILR-beta | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
| activating receptor PILRbeta, Activating receptor PILR-beta, Cell surface receptor FDFACT, cell surface receptor FDFACT1, cell surface receptor FDFACT2, FDFACT, FDFACT1, FDFACT2, paired immunoglobin-like receptor beta, paired immunoglobin-like type 2 receptor beta, paired immunoglobulin-like receptor beta, paired immunoglobulin-like type 2 receptor beta | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PILRB | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AGHPEIGEAAVAVHQGDQTHHHPGCHNHHHLEAQQHNHHSRPQGHRKQRALRIMAPKSGHCHQGCIGCRCAQNCHFGTAVPPPPVVEEKER | |
| 0.1 mL | |
| Signal Transduction | |
| 29990 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction