Learn More
Invitrogen™ PIK3CD/PIK3R1 (p110 delta/p85 alpha) Recombinant Human Protein, His tag
Phosphatidylinositol 3-kinase (PI3K) is composed of a catalytic subunit (p110δ) and a regulatory subunit (p85α in this case), which are encoded by the PIK3CD and PIK3R1 genes. PI3K phosphorylates phosphoinositides on the 3-hydroxyl group of the inositol ring.
Brand: Invitrogen™ PV6452
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Phosphatidylinositol 3-kinase (PI3K) plays a role in oncogenic transformation and is consistently expressed at a high level in AML blast cells. PI3K can play a selective role in the amplification of phosphatidylinositol (3,4,5) trisphosphate (PIP3) levels that leads to neutrophil polarization, and regulates directional migration, but not random locomotion. The PI3K signaling pathway is involved in immune responses relevant to the pathogenesis of rheumatoid arthritis and other inflammatory diseases.
- Available in 3 convenient sizes
- Validated in our Adapta™ and Kinase Binding assays
- A full length protein that can help advance your research when used with validated assays
Specifications
NP_852664.1/NP_005017.3 | |
50mM sodium phosphate (pH 7.0), 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, and 25% glycerol. | |
120.6kDa/84.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
p110 delta/p85 alpha/regulatory subunit 1 (alpha)/p110δ/p85α/phosphoinositide-3-kinase | |
PI3 Kinase Family | |
Recombinant | |
Baculovirus |
Kinase Assay, Industrial and Applied Science, Kinase Biology, Pharma and Biopharma, Target and Lead Identification and Validation | |
5295, 5293 | |
100μg | |
Full Length | |
PIK3R1/PIK3CD | |
Human | |
His Tag |