Learn More
Invitrogen™ PIK3CD/PIK3R1 (p110 delta/p85 alpha) Recombinant Human Protein, His tag
Phosphatidylinositol 3-kinase (PI3K) is composed of a catalytic subunit (p110δ) and a regulatory subunit (p85α in this case), which are encoded by the PIK3CD and PIK3R1 genes. PI3K phosphorylates phosphoinositides on the 3-hydroxyl group of the inositol ring.
£536.00 - £6690.00
Specifications
KinaseFamily | PI3 Kinase Family |
---|---|
Species | Human |
Recombinant | Recombinant |
Protein Tag | His Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15429063
|
Invitrogen™
PV6451 |
20 μg |
£536.00
20µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15449063
|
Invitrogen™
PV6452 |
100μg |
£1554.00
100µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15459063
|
Invitrogen™
PV6453 |
1mg |
£6690.00
1mg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Phosphatidylinositol 3-kinase (PI3K) plays a role in oncogenic transformation and is consistently expressed at a high level in AML blast cells. PI3K can play a selective role in the amplification of phosphatidylinositol (3,4,5) trisphosphate (PIP3) levels that leads to neutrophil polarization, and regulates directional migration, but not random locomotion. The PI3K signaling pathway is involved in immune responses relevant to the pathogenesis of rheumatoid arthritis and other inflammatory diseases.
- Available in 3 convenient sizes
- Validated in our Adapta™ and Kinase Binding assays
- A full length protein that can help advance your research when used with validated assays
Specifications
PI3 Kinase Family | |
Recombinant | |
Baculovirus |
Human | |
His Tag |