missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PIG3 Antibody (10A2), Novus Biologicals™
Mouse Monoclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | PIG3 |
|---|---|
| Clone | 10A2 |
| Dilution | Western Blot : 5-10 μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066967
|
Novus Biologicals™
NBP3-30804-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067134
|
Novus Biologicals™
NBP3-30804-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PIG3 Monoclonal antibody specifically detects PIG3 in Human samples. It is validated for Western BlotSpecifications
| PIG3 | |
| Western Blot : 5-10 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| p53-induced gene 3 protein, PIG3quinone oxidoreductase homolog, quinone oxidoreductase PIG3, tumor protein p53 inducible protein 3, Tumor protein p53-inducible protein 3 | |
| Hybridoma produced by the fusion of splenocytes from mice immunized with recombinant human PIG3 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 10A2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis | |
| Phosphate buffered saline | |
| 9540 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title