Learn More
Novus Biologicals™ PIG3 Antibody (10A2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30804-25ug
 View more versions of this product
Product Code. 30066967
  • £162.00 / 25µg
Description
PIG3 Monoclonal antibody specifically detects PIG3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PIG3
Monoclonal
Unconjugated
Phosphate buffered saline
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Western Blot
10A2
Western Blot : 5-10 μg/mL
p53-induced gene 3 protein, PIG3quinone oxidoreductase homolog, quinone oxidoreductase PIG3, tumor protein p53 inducible protein 3, Tumor protein p53-inducible protein 3
Hybridoma produced by the fusion of splenocytes from mice immunized with recombinant human PIG3 protein.
25 μg
Apoptosis
9540
Store at -20°C. Avoid freeze/thaw cycles.
IgG1