missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ NHS-Rhodamine Antibody Labeling Kit

Product Code. 10560054 Shop All Thermo Scientific Products
Click to view available options
Quantity:
3 Reactions
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10560054

Brand: Thermo Scientific™ 53031

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Label antibodies, proteins and other molecules for fluorescence imaging (ex 546; em 572-615) using these activated derivatives of red-fluorescent rhodamine dye.

Thermo Scientific™ TRITC and Rhodamine Labeling Reagents and Kits are high-performance derivatives of rhodamine dye, activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes.

Rhodamine dyes are used in fluorescence microscopy, flow cytometry and immunofluorescence-based assays such as Western blotting and ELISA. TRITC, NHS-rhodamine and Texas Red™ Sulfonyl Chloride are reactive towards primary amine groups on proteins, peptides and other biomolecules. Rhodamine dyes are often a mixture of isomers with reactive groups attached at the 5- and 6-positions of the bottom ring. The properties of these isomers are indistinguishable in terms of excitation and emission spectra and for protein applications there is no need to isolate a specific isomer. The single isomer is used in high resolution separation techniques such as sequencing and capillary electrophoresis.

Highlights:

  • Label and purify antibody in about one hour Efficiently labels antibodies and other purified proteins at primary amines (lysine side chains) Standard protocol results in antibodies with excellent dye:protein ratios for optimum activity and fluorescence Single-use fluors: no need to weigh tiny amounts powder; kits contain single-use vials of reagent Efficient purification and removal of non-reacted dye with excellent protein recovery

Includes:

NHS-Rhodamine Antibody Labeling Kit (Mfr. No. 53031)

NHS-Rhodamine, 3 vials; Borate Buffer, 1mL; Purification Resin, 5mL; Spin Columns, 6 each; Collection Tubes, 12 each

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce Rhodamine Antibody Labeling Kit
Product Type Antibody Labeling Kit
Quantity 3 Reactions
Sufficient For Labeling and purifying 3 x 1 mg (2 mg/mL) of IgG or other proteins
Content And Storage Sufficient For: Labeling and purifying 3 x 1 mg (2 mg/mL) of IgG or other proteins
  • NHS Rhodamine, 3 x 35 μg vials
  • Borate Buffer, 1 mL
  • Purification Resin, 5 mL
  • Spin Columns, 6 each
  • Collection Tubes, 12 each

    Store NHS-Rhodamine at -20°C. Store all other components at 4°C.
  • Excitation/Emission 552/575
    Labeling Scale 0.5 to 1 mg
    Product Line Pierce™
    Label or Dye Rhodamine Isomers
    Labeling Target Antibodies, Proteins
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.