missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Thermo Scientific™ Pierce Traut's Reagent is 2-iminothiolane, a small thiolation compound that reacts with primary amines (e.g., lysine side chains) to add a small spacer arm (8.1 Ås) terminated by a free sulfhydryl group.
Highlights:
- Reacts with primary amines (lysine epsilon amino groups) at pH 7-9
- Adds a short spacer ending in a sulfhydryl group, ready for use as a target for sulfhydryl-specific crosslinking or labeling
- Amine-containing spacer arm preserves charge of original, modified primary amine
Specifications
Specifications
| Chemical Reactivity | Amine |
| Content And Storage | Storage: Upon receipt store product at 4°C with desiccant. Product shipped at ambient temperature |
| Label or Dye | Sulfhydryl Group |
| Label Type | Conjugating Groups |
| Product Line | Pierce |
| Spacer | Short (<10 Å) |
| Form | Powder |
| Quantity | 500 mg |
| Solubility | DMF, DMSO |
| Product Type | Thiophilic Adsorption Kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?