missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ TCEP-HCl, No-Weigh™ Format

Product Code. 11856784 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 1 mg
1 g
10 g
Unit Size:
10g
1g
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11856784 10 g 10g
10530434 1 g 1g
15780329 10 x 1 mg Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11856784 Supplier Thermo Scientific™ Supplier No. 20491

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reduce protein and peptide disulfide bonds using this thiol-free, pure, crystalline tris (2-carboxyethyl) phosphine hydrochloride, TCEP (CAS 5961-85-3).

Thermo Scientific™ Pierce TCEP-HCl is pure, crystalline Tris(2-carboxyethyl)phosphine hydrochloride, a thiol-free compound that is highly effective at reducing protein and peptide disulfide bonds, as well as being odorless and stable.

Highlights:

  • Reduces even the most stable water-soluble alkyl disulfides
  • Effective at room temperature and pH 5 in less than five minutes
  • Does not require removal before reaction with maleimide or haloacetyl compounds
  • Water solubility of 310g/L
  • Resistant to air oxidation; nonvolatile and nonreactive toward other functional groups in proteins
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store at room temperature in sealed container to prevent oxidation. Product shipped at ambient temperature.
Product Line Pierce
Form Solid
Quantity 10 g
Solubility Water
Product Type Disulfide Reducing Reagent
compliance-icons
Product Identifier
  • TCEP HCl, 10 g.Tris{ 2-carboxyethyl} phosphine
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Tris(2-carboxyethyl)phosphonium chloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.