missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-SHPP (Water-Soluble Bolton-Hunter Reagent)

Product Code. 11801435 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
11801435 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 11801435

Brand: Thermo Scientific™ 27712

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Add tyrosine (tyrosyl) groups to proteins to increase the number of iodine-125 labeling (iodination) sites using an amine-reactive, NHS ester compound.

Thermo Scientific™ Pierce Sulfo-SHPP (water-soluble Bolton-Hunter Reagent) conjugates tyrosine-like functional groups to primary amines to increase the number of tyrosyl groups that can be iodinated by iodine-125 labeling procedures.

Highlights:

  • Membrane-impermeable
  • Ideal for selectively radiolabeling cell surface proteins
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sulfo-SHPP
Quantity 100 mg
Content And Storage Upon receipt store at -20°C. Product is shipped at ambient temperature.
Product Line Pierce
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Iodine-125
Spacer Short
Label Type Radioisotope
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.