missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Thermo Scientific™ Pierce Sulfhydryl Addition Kit is a complete set of reagents, including SATA, to enable modification of primary amines to free sulfhydryl groups for use in heterobifunctional crosslinking techniques.
Highlights:
- Reacts with primary amines to add protected sulfhydryls
- An easy deprotection step generates a free sulfhydryl group
- Thermo Scientific™ Sulfhydryl Addition Kit contains all necessary components for thiolation, deprotection, purification and quantitation
Specifications
Specifications
| Chemical Reactivity | Amine |
| Content And Storage | Sufficient For: One modification experiment with 0.5 to 5 mg of purified protein • SATA, 2 mg • Hydroxylamine·HCl, 5 mg • Conjugation Buffer Stock (10X), 20 mL • PBS pack (makes 500 mL), 1 pack • Dimethylformamide (DMF), 1 mL • Dextran Desalting Column, 1 x 5 mL • Column Extender, 1 • Ellman's Reagent (DTNB), 2 mg • Cysteine·HCl, 20 mg Store at 4°C. |
| Label or Dye | Sulfhydryl Group |
| Label Type | Conjugating Groups |
| Product Line | Pierce |
| Crosslinker Type | Heterobifunctional |
| Reactive Moiety | Active Ester, Succinimidyl Ester, NHS Ester |
| Spacer | Short (<10 Å) |
| Form | Powder |
| Reagent Type | SATA |
| Show More |
Safety and Handling
Product Identifier
- Pierce Sulfhydryl Addition Kit
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: disodium hydrogenorthophosphate, L-Cysteine, hydrochloride,3,3'-dithiobis [6-nitrobenzoic] acid, N,
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?