missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SDS-Out™ SDS Precipitation Kit

Product Code. 10341554 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10341554 10 mL kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 10341554 Supplier Thermo Scientific™ Supplier No. 20308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

Quickly remove SDS from small-volume (0.1 to 10mL) protein samples with this SDS precipitation kit.

Perform convenient removal of SDS from small-volume (0.1 to 10mL) protein samples with Thermo Scientific™ SDS-Out SDS Precipitation Reagent.

The simple reagent and kit effectively remove excess SDS from small protein samples. Sodium dodecyl sulfate (SDS) is a detergent typically used in cell lysis reagents and other protein solubilization buffers. Traditional methods for SDS removal include prolonged dialysis, anion exchange chromatography, affinity columns or and acetone precipitation. By comparison, the SDS-Out Precipitation Reagent is relatively easy to use and provides excellent protein recovery.

Highlights:

  • Convenient removal of SDS from microliter-volume samples
  • Use for samples containing 0.1 to 1% SDS
  • Minimal dilution of small-volume samples
  • Protein recovery is 90 to 100% (based on tests with BSA, cytochrome C, soybean trypsin inhibitor, ovalbumin, ribonuclease A, myoglobin and human IgG)

Includes:

Spin cup and collection tube accessories for use with the SDS-Out Reagent

TRUSTED_SUSTAINABILITY

Specifications

Product Type SDS Precipitation Kit
Content And Storage Sufficient For: Precipitation of SDS from 200 mL of protein solution
• SDS Precipitation Reagent, 10 mL
• Spin Cup Columns, 12 columns
• Collection Tubes, 12 tubes

Upon receipt, store components at 4°C.
Column Type Precipitation, Filtration, n/a
Product Line SDS-Out
Purification Target Detergent Removal
Sufficient For 200 mL Protein Solution
Quantity 10 mL kit
Format Kit, Microcentrifuge, Spin Column

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.