missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Premium Grade TCEP-HCl

Product Code. 15295763 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15295763 1 g 1g
15225773 10 g 10g
15235773 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15295763 Supplier Thermo Scientific™ Supplier No. PG82080

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this disulfide reducing agent.

Optimize applications where product integrity and risk minimization are paramount with the specially characterized Thermo Scientific™ Pierce™ Premium Grade TCEP-HCl, our highest quality formulation of this disulfide reducing agent.

Thermo Scientific Pierce Premium Grade TCEP-HCl is our highest quality formulation of this disulfide reducing agent, specially characterized for applications where product integrity and risk minimization are paramount.

Compared to the standard grade product, Premium Grade SPDP provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. TCEP-HCl is pure, crystalline Tris(2-carboxyethyl)phosphine hydrochloride, a thiol-free compound that is highly effective at reducing protein and peptide disulfide bonds, as well as being odorless and stable. TCEP selectively and completely reduces even the most stable water-soluble alkyl disulfides over a wide pH range.

Highlights:

  • TCEP – a thiol-free, odorless yet potent reducing agent for cleaving disulfide bonds
  • High quality – identity and purity confirmed by several tests, including quantitative NMR
  • Product integrity – enhanced level of testing and characterization compared to standard grade
  • Lot retention – ample supply of past lots retained to ensure future process testing
  • Change management – Change Control Notification (CCN) service
  • Consistent manufacture – batch-specific manufacturing documentation review

Advantages of TCEP-HCl over other reducing agents:

  • Odorless – Unlike DTT or BME, TCEP is odor-free, so reductions can be carried out conveniently on the bench top.
  • Stable in air – The inherent stability of the TCEP moiety eliminates the need for any special precautions to avoid oxidation when handling, using or storing TCEP.
  • Efficient – For most applications, 5 to 50mM TCEP provides sufficient molar excess to effectively reduce peptide or protein disulfide bonds within a few minutes at room temperature.
  • Compatible – With TCEP, removal of the reducing agent is not necessary prior to most applications, (e.g. histidine-tagged protein purification, maleimide conjugations).

Stability in Solution
TCEP is stable in aqueous, acidic and basic solutions. When TCEP is dissolved directly in water, the resulting pH is approximately 2.5. Studies indicate that no change in concentration of TCEP occurs after 24-hour incubation at room temperature in 100mM HCl, 100mM NaOH, or in any of the following 50mM buffers: Tris·HCl (pH 7.5, 8.5 and 9.5), HEPES (pH 6.8 and 8.2), borate (pH 8.2 and 10.2) and CAPS (pH 9.7 and 11.1).5 Even after three weeks in these buffers, less than 20% of the TCEP was oxidized.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store product at room temperature in sealed container to prevent oxidation. Product is shipped at ambient temperature.
Product Line Pierce
Form Solid
Quantity 1 g
Product Type Disulfide Reducing Reagent
compliance-icons
Product Identifier
  • Pierce Premium Grade, TCEP.HCl
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Propanoic acid, 3,3',3''- phosphinidynetris-, hydrochloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.