Learn More
Thermo Scientific™ MA(PEG)12 Methyl-PEG-Amine Compound
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce MA(PEG)n Compounds are methyl- and amine-terminated polyethylene glycol reagents, where n equals 4 to 24 PEG units, that are useful for a variety of surface-modification and molecule-pegylation applications.
MA(PEG)n compounds have the form Methyl-PEGn-Amine and are ideal for derivatizing surfaces and molecules with discrete numbers (n = 4, 8, 12, 24) of ethylene glycol groups. Use any amine-reactive chemistry (e.g., EDC or NHS ester) to PEGylate surfaces, proteins, or other molecules with a precise number of hydrophilic, nonimmunogenic PEG units.
Highlights:
- Pure compounds of specific size ensure complete control of PEGylation
- Straight-chain compounds with simple terminal primary amine
- Adaptable to a variety of modification procedures
Recommended for:
- PEGylate carboxylate or amine surfaces Add inert mass to proteins, immunogens, drug compounds and probes Improve solubility (decrease aggregation) of proteins or peptides without affecting function Protect proteins from proteolysis
Specifications
Specifications
| Chemical Reactivity | Aldehyde |
| Content And Storage | Upon receipt store product at -20°C protected from moisture. Product is shipped at ambient temperature. |
| Label Type | Polyethylene Glycol (PEG), Blocking Groups |
| Reactive Moiety | Amine, Primary Amine |
| Spacer | 12 ethylene glycol units |
| Form | Solid |
| Quantity | 100 mg |
| Solubility | Water |
| Product Type | Amine Compound |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.