missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MA(PEG)12 Methyl-PEG-Amine Compound

Product Code. 10230444 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 10230444

Brand: Thermo Scientific™ 26115

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MA(PEG)n: methyl- and amine-capped polyethylene glycol compounds containing 4 to 24 PEG units; use for surface-modification and molecule-pegylation applications.

Thermo Scientific™ Pierce MA(PEG)n Compounds are methyl- and amine-terminated polyethylene glycol reagents, where n equals 4 to 24 PEG units, that are useful for a variety of surface-modification and molecule-pegylation applications.

MA(PEG)n compounds have the form Methyl-PEGn-Amine and are ideal for derivatizing surfaces and molecules with discrete numbers (n = 4, 8, 12, 24) of ethylene glycol groups. Use any amine-reactive chemistry (e.g., EDC or NHS ester) to PEGylate surfaces, proteins, or other molecules with a precise number of hydrophilic, nonimmunogenic PEG units.

Highlights:

  • Pure compounds of specific size ensure complete control of PEGylation
  • Straight-chain compounds with simple terminal primary amine
  • Adaptable to a variety of modification procedures

Recommended for:

  • PEGylate carboxylate or amine surfaces Add inert mass to proteins, immunogens, drug compounds and probes Improve solubility (decrease aggregation) of proteins or peptides without affecting function Protect proteins from proteolysis
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Aldehyde
Content And Storage Upon receipt store product at -20°C protected from moisture. Product is shipped at ambient temperature.
Label Type Polyethylene Glycol (PEG), Blocking Groups
Reactive Moiety Amine, Primary Amine
Spacer 12 ethylene glycol units
Form Solid
Quantity 1 g
Solubility Water
Product Type Amine Compound

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.