missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mammalian beta-Galactosidase Assay Kit

Product Code. 10561094 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
75 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10561094 75 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10561094 Supplier Thermo Scientific™ Supplier No. 75707
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lyse cultured mammalian cells and measure beta-galactosidase activity using compatible M-PER Cell Lysis Reagent and colorimetric b-Gal Assay Reagent.

The Thermo Scientific Mammalian β-Galactosidase Assay Kit provides a simple colorimetric method for lysing cultured mammalian cells and measuring beta-Gal activity.

Features of the Mammalian β-Galactosidase Assay Kit:

  • Choose stand-alone assay reagent or complete kit for cell lysis and detection
  • Colorimetric assay easily measured in a plate reader or spectrophotometer (405nm)
  • Adaptable to 96-well microplates or tube assay formats
  • Perform homogeneous screening assays or measure activity secondarily after cell lysis
  • Also compatible with prepared lysates from other species such as bacteria

The Assay Reagent and Kit comprise a simple and versatile assay system for measuring β-galactosidase activity. Mammalian cells grown on a plate may be lysed and tested for β-galactosidase activity by adding a single, all-in-one lysis-and-assay solution. Alternatively, the included Thermo Scientific M-PER Mammalian Protein Extraction Reagent may be used to prepare lysate from washed adherent or pelleted cells, and then a portion of the lysate used to quantitate the β-Gal activity. With M-PER Reagent, there is no need to scrape cells or to perform freeze-thaw cycles. The remaining lysates prepared with M-PER Reagent may be used for applications such as reporter assays (luciferase, β-galactosidase and chloramphenical acetyltransferase), protein kinase assays (PKA, PKC, and Tyrosine Kinase), immunoassays (Western blot, ELISA, RIA) and protein purification.

Related Products

beta-Galactosidase Assay Reagent (Cat. No. 75705)

beta-Galactosidase Assay Stop Solution (Cat. No. 75706)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: 250 microplate wells of adherent cells
  • Pierce beta-Galactosidase Assay Reagent, 25 mL
  • M-PER Mammalian Protein Extraction Reagent, 25 mL
  • Stop Solution, 25 mL

    Store the β-Galactosidase Assay Reagent at -20°C. Store all other components at room temperature.
  • Description Pierce Mammalian beta-Galactosidase Assay Kit
    Detection Method Absorbance
    For Use With (Application) Absorbance
    For Use With (Equipment) Microplate Reader
    Product Type Beta-Galactosidase Assay Kit
    Dye Type Enzyme Labeled
    Format 96-well Plate
    Product Line Pierce
    Quantity 75 mL
    Substrate Proprietary
    Substrate Type Beta-Gal Substrate
    Sufficient For 250 microplate wells of adherent cells
    Target Beta-Galactosidase
    Absorbance Wavelength 405 nm
    Show More Show Less
    compliance-icons
    Product Identifier
    • Mammalian beta-Galactosidase Assay Kit
    Signal Word
    • Warning
    Hazard Category
    • Serious eye damage/eye irritation Category 2
    Hazard Statement
    • H319-Causes serious eye irritation.
    Precautionary Statement
    • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
    • P264-Wash hands thoroughly after handling.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Supplemental information
    • MIXTURE LIST-Contain: sodium carbonate

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.