missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Iodoacetic Acid

Product Code. 11815005 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
24 x 9.3 mg
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11815005 24 x 9.3 mg Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11815005 Supplier Thermo Scientific™ Supplier No. 90034

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Get rapid carboxymethylation of reduced cysteine residues with Thermo Scientific™ Pierce™ Iodoacetic Acid and Iodoacetamide.

Thermo Scientific™ MALDI Matrices are highly purified, recrystallized reagents packaged in convenient single-use tubes sufficient for analyzing up to 96 samples by matrix assisted laser desorption ionization.

Iodoacetic Acid (IAA) can react with several protein functional groups but is typically used for specific S-carboxymethylation of sulfhydryls (reduced cysteines).

Iodoacetic acid reacts with sulfhydryls on cysteines, imidazolyl side chain nitrogens of histidines, the thioether of methionine and the primary amine group of lysines.The rate of reaction and specificity is dependent on the ionization level, which can be manipulated by the pH of the reaction condition.

Highlights:
  • React at slightly alkaline pH for specific S-carboxymethylation of free sulfhydryls
  • React at low pH for specific carboxymethylation of methionines
  • React at high pH to favor carboxymethylation of histidines and lysines
  • Methylate reduced cysteine peptide fragments in protease digests for mass spectrometry
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Sulfhydryl
Description Iodoacetamide, Single-Use
Formulation White solid of 2-iodoacetamide (IAM) in foil-sealed black microtubes (3 x 8 format)
Molecular Weight (g/mol) 184.96
Sufficient For 24 separate uses for alkylation before mass spectrometry, each with up to 75 protein samples
Quantity 24 x 9.3 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.