missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ActivX™ Desthiobiotin-ATP Probe

Product Code. 11822134 Shop All Thermo Scientific Products
Click to view available options
Quantity:
16 x 12.6 μg
Unit Size:
Pack of 16
This item is not returnable. View return policy

Product Code. 11822134

Brand: Thermo Scientific™ 88311

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ActivX™ probes have been designed to selectively label and enrich kinases through binding of their respective active sites with desthiobiotin tagged ATP or ADP to enable assessment of enzyme activity.

ActivX Desthiobiotin-ATP or ADP Probes are nucleotide derivatives that covalently modify the active site of enzymes at conserved lysine residues in the nucleotide binding site. The structure of these probes consists of a modified biotin (desthiobiotin) attached to the nucleotide through a labile acyl-phosphate bond. Desthiobiotin is a biotin analog that binds less tightly to biotin-binding proteins resulting in binding that is easily reversed by biotin displacement, low pH or heat denaturation.

ActivX active-site enrichment probes are advantageous for determining active enzyme levels using analog-tagged probes to allow profiling of both inactive and active enzymes. The ActivX ATP and ADP Probes covalently label the active site of ATPases, including chaperones and metabolic enzymes, to enable their selective enrichment using a desthiobiotin tag.

Desthiobiotin-ATP or ADP probes can be used to selectively enrich, identify and profile target enzyme classes or assess the specificity of enzyme inhibitors. Because many ATPases and other nucleotide-binding proteins bind nucleotides or inhibitors even when they are enzymatically inactive, the desthiobiotin probes allow profiling of both inactive and active enzymes in a complex sample. Preincubation of samples with small-molecule inhibitors that compete for active sites can be used to determine inhibitor binding affinity. Active-site nucleotide probes also can be used to identify inhibitor off-targets. With selective enrichment, one can identify and profile target enzyme classes across samples or can assess the specificity and affinity of enzyme inhibitors.

Applications of ActivX ATP and ADP Probes:
• Profile small-molecule binding affinities and active-site inhibition in a dose-dependent manner
• Identify dozens to hundreds of inhibitor targets and off-targets from tissues, cells and subcellular proteomes
• Enrichment of enzymes based on function

Assessment of active-site labeling with desthiobioin probes can be accomplished by either Western blot or mass spectrometry (MS). For the Western blot workflow, desthiobiotin-labeled proteins are enriched for SDS-PAGE analysis and subsequent detection with specific antibodies. For the MS workflow, desthiobiotin-labeled proteins are reduced, alkylated and enzymatically digested to peptides. Only the desthiobiotin-labeled, active-site peptides are enriched for analysis by LC-MS/MS. Both workflows can be used for determining inhibitor target binding, but only the MS workflow can identify global inhibitor targets and off-targets.

These products are subject to a limited use label license.

TRUSTED_SUSTAINABILITY

Specifications

Description ActivX™ Desthiobiotin-ATP Probe
Formulation Desthiobiotin-ATP, MW 1259
Content And Storage Upon receipt store at –80°C.
Assay Protein Quantitation Assay
Detection Method Chemiluminescence, Mass Spectrometry, Western Blot
Substrate Type Kinase Substrate
Product Line ActivX
Quantity 16 x 12.6 μg
For Use With (Application) Pull-Down
Target Kinases (Multiple), Phosphodiesterases, ATPases, GTPases, Phosphatases
Type Labeled Nucleotide Probe
Label or Dye Biotin and Other Haptens
Format Tube, Spin Column
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.