missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Iodination Beads
Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 Beads
Unit Size:
Pack of 50
Description
Thermo Scientific™ Pierce Iodination Beads are 3mm diameter polystyrene beads that are coated with an oxidizing reagent, which provides efficient activation of iodine-125 for protein or peptide iodination procedures.
Highlights:
- Derivatized, uniform, nonporous polystyrene beads
- Radioiodide incorporation as high as 99%; labeled protein recovery >90%
- Functions over a broad pH and temperature range
- Fast and easy to use: iodinations complete in 2 to 15 minutes
- Reaction stopped by simply removing beads from reaction mixture with tweezers or pasteur pipet; no reducing agent necessary to terminate reaction
Additional Features:
- Iodinates in the presence of azides, detergents, urea and high salt
- Allows efficient iodination of cell membrane surface proteins
- More gentle method for iodination than soluble chloramine-T because there is no contact between the protein and the immobilized oxidizing agent1
- Highly reproducible iodinations
- Can be used to quantitatively iodinate histidine at pH 8.22
Specifications
Specifications
| Product Type | Iodination Bead |
| Quantity | 50 Beads |
| Content And Storage | Upon receipt store product desiccated at 4°C. Avoid exposing beads to moisture or reducing agents. Product is shipped at ambient temperature. |
| Product Line | Pierce |
| Label or Dye | Iodine-125 |
| Label Type | Radioisotope |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction