Learn More
Description
Perform immunoprecipitation (IP) assays of HA-tagged proteins or co-IP reactions with HA-tagged bait proteins with the magnetic beads, positive control and reagents provided in the Thermo Scientific™ Pierce Magnetic IP/Co-IP Kit.
Additional Information and Online Ordering
The complete Magnetic IP/Co-IP Kit can be used to perform 40 IP or Co-IP assays. It includes magnetic beads coated with immobilized anti-HA antibody, lysis/wash buffer, low pH elution buffer, neutralization buffer, HA-tag positive control lysate, and non-reducing sample buffer. Protocols are provided for both manual and automated workflows.
Highlights:
- Specific – highly specific anti-HA monoclonal antibody enables high yields of immunoprecipitation products
- Fast – Optimized protocol is completed in approximately 1 hour
- Low non-specific binding – stable, pre-blocked beads provide highly purified HA-tagged proteins
- Versatile – beads are compatible with manual and automated workflows (e.g., Thermo Scientific™ KingFisher Instruments)
- Convenient and easy – complete kit includes all necessary reagents and easy-to-follow instructions
Includes:
The kit contains magnetic beads immobilized with anti-HA antibody, lysis/wash buffer, low pH elution buffer, neutralization buffer, HA-tag positive control lysate, and non-reducing sample buffer.
Recommended for:
High-throughput immunoprecipitation of recombinant HA-tagged proteins and co-IP of interacting proteins
Specifications
Specifications
| Capacity (Metric) | 1 mL |
| Formulation | Slurry: 10 mg/mL, 1% solids |
| Content And Storage | Sufficient For: 40 IP reactions using 25 μL of anti-HA magnetic beads • HA-tagged Positive Control (Part No. 26180X), 500 μL • Pierce Anti-HA Magnetic Beads, 1 mL • Pierce IP Lysis/Wash Buffer, 2 x 50 mL • Non-Reducing Sample Buffer (5X), 5 mL • Elution Buffer (pH 2.0), 5 mL • Neutralization Buffer (pH 8.5), 1 mL HA-tagged Positive Control ships separately—store at -70°C. Store remaining components at 4°C. |
| Assay | Affinity Purification |
| For Use With (Equipment) | KingFisher™ Flex, Magnetic Stand |
| Product Line | Pierce |
| Quantity | 40 reactions |
| For Use With (Application) | Pull-Down (Tag-Based), Immunoprecipitation (IP) |
| Target | Target-Specific |
| Type | Tag IP/Co-IP Kit |
| Show More |
Safety and Handling
- Pierce HA-Tag Magnetic IP/CoIP Kit
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: hydrogen chloride
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.