missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ NHS-Fluorescein Antibody Labeling Kit

Product Code. 10628664 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3 reactions kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10628664 3 reactions kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10628664 Supplier Thermo Scientific™ Supplier No. 53029
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Activated derivatives of green-fluorescent fluorescein dye to label antibodies, proteins and other molecules for fluorescence imaging (ex 494; em 518).

Thermo Scientific™ FITC and Fluorescein Labeling Reagents and Kits are high-performance derivatives of fluorescein dye, activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes.

Fluorescein dyes are used in wide-ranging applications including fluorescence microscopy, flow cytometry and immunofluorescence-based assays such as Western blotting and ELISA. FITC and NHS-fluorescein are reactive toward primary amine groups on proteins, peptides and other biomolecules, while fluorescein-5-maleimide and 5-IAF react with free sulfhydryls on cysteine residues.Most activated fluorescein derivatives are mixture of isomers with reactive groups attached at the 5- and 6-positions of the bottom ring. The properties of these isomers are indistinguishable in terms of excitation and emission spectra and for protein applications there is no need to isolate a specific isomer. The single isomer is used in high resolution separation techniques such as sequencing and capillary electrophoresis.

Highlights:

  • Amine-specific labeling: the NHS-ester or isothiocyanate efficiently labels antibodies and other purified proteins at primary amines (lysine side chains) Optimized procedure: following the standard protocol results in antibodies with excellent dye:protein ratios for optimum activity and fluorescence Single-use fluors: no need to weigh tiny amounts powder; kits contain single-use vials of reagent Efficient purification: kits include purification resin and easy-to-use spin columns, ensuring rapid and efficient removal of non-reacted dye and excellent protein recovery
TRUSTED_SUSTAINABILITY

Specifications

Description Pierce Fluorescein Antibody Labeling Kit
Product Type Antibody Labeling Kit
Quantity 3 reactions kit
Sufficient For Labeling and purifying 3 x 1 mg (2 mg/mL) of IgG or other proteins
Content And Storage Sufficient For: Labeling and purifying 3 x 1 mg (2 mg/mL) of IgG or other proteins
  • NHS Fluorescein, 3 x 50 μg vials
  • Borate Buffer, 1 mL
  • Purification Resin, 5 mL
  • Spin Columns, 6 each
  • Collection Tubes, 12 each

    Store NHS-Fluorescein at -20°C. Store other kit components at 4°C.
  • Excitation/Emission 494/518
    Labeling Scale 0.5 to 1 mg
    Product Line Pierce
    Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
    Label or Dye FITC (Fluorescein)
    Labeling Target Antibodies, Proteins
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.