missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Dye Removal Columns

Product Code. 10483415 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 columns, 5 mL resin
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10483415 10 columns, 5 mL resin Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10483415 Supplier Thermo Scientific™ Supplier No. 22858
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Affinity resin with spin columns to quickly bind and remove excess fluorescent dye from antibody (protein) solutions following labeling reactions.

Thermo Scientific™ Pierce Dye Removal Columns effectively bind to unconjugated fluorescent dyes molecules from protein solutions to rapidly purify fluorescent conjugated antibodies and other proteins after labeling reactions.

Highlights:

  • Fast - removal free dye from conjugates in approximately 30 seconds
  • Efficient - purification resin provides outstanding conjugate recovery
  • Minimal sample dilution
TRUSTED_SUSTAINABILITY

Specifications

Format Kit, Microcentrifuge, Spin Column
Quantity 10 columns, 5 mL resin
Product Type Dye Removal Column
Content And Storage Store purification resin at 4°C. Shipped at ambient temperature.
Target Fluorescent Dye Removal
Product Line Pierce

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.