missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ DTT (Dithiothreitol)

Product Code. 11806754 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
48 x 7.7 mg
Unit Size:
0.1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11806754 48 x 7.7 mg 0.1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11806754 Supplier Thermo Scientific™ Supplier No. 20291

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ Pierce™ DTT (Dithiothreitol) No-Weigh™ Format is designed for convenient preparation of stocks that will be added to protein samples for effective reduction of disulfide bonds.

Thermo Scientific™ Pierce DTT (Dithiothreitol) is offered as 7.7 mg x 48-tube microplates for convenient preparation of 100 μL of 0.5 M stocks for addition to protein samples for effective reduction of disulfide bonds.

Thermo Scientific Pierce DTT (Dithiothreitol), No-Weigh format, is offered as 7.7 mg x 48-tube microplates for convenient preparation of 100 μL of 0.5 M single-use stocks for addition to protein samples for effective reduction of disulfide bonds.

DTT is a popular reducing agent for many protein research applications, including preparing proteins for reducing polyacrylamide electrophoresis (SDS-PAGE) and reducing disulfide bonds for labeling sulfhydryl groups in conjugation reactions. At low concentrations, DTT is used to stabilize proteins and enzymes with free sulfhydryl groups to keep monothiols in a reduced state. DTT reduction is also used to prepare proteins for mass spectrometry.

Properties of DTT
Synonyms: 1,4-dithiothreitol; DTT; Cleland’s Reagent; 1,4-dimercapto-2,3-butanediol
Molecular weight (MW): 154.24
Molecular formula: C4H10O2S2
CAS number: 3483-12-3

DTT is not stable in solution and should be prepared fresh for each use. The No-Weigh format consists of a 48-microtube tray containing dry, stable, single-use aliquots of the DTT chemical to eliminate the need to weigh small amounts of reagent and prevent waste of unused DTT. Just puncture the seal of one microtube, add 100 μL of water or buffer, and mix to dissolve. In just a few seconds you will have a solution of 500 mM DTT, ready for a variety of laboratory methods. Dilute as necessary or add the appropriate volume of the 500 mM stock solution to achieve the amount of DTT reducing agent required for your specific application.

Features of the No-Weigh format
Saves time—just add 100 μL water or buffer to make 500 mM DTT solution in seconds
Eliminates waste—makes 100 μL at a time, just the right amount for most applications
Always fresh—ensures full reducing strength for every use; no need to store stock solutions

With the convenient No-Weigh format, a 500 mM DTT solution can be made in less than 30 seconds. The unique packaging ensures that the DTT reducing agent is at full strength and able to protect proteins from oxidative damage or reduce any disulfides prior to electrophoresis.

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce DTT, No-Weigh Format
Formulation Pure (approx. 98%) white powder of dithiothreitol
Sufficient For Preparing 100 μL of 0.5M DTT per microtube
Quantity 48 x 7.7 mg
compliance-icons
Product Identifier
  • PIERCE DTT, NO-WEIGH
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: 2, 3-Butanediol, 1,4-dimercapto-, (R, R)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.