missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Cysteine-HCl

Product Code. 11889350 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11889350 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11889350 Supplier Thermo Scientific™ Supplier No. 44889
  • ONLINE EXCLUSIVE PRICE
    £89.00
    £79.65 / 5g
    Save £9.35
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Make sulfhydryl assay standards or perform protein refolding experiments using this pure, crystalline cysteine hydrochloride (Cys-HCl).

Thermo Scientific™ Pierce Cysteine-HCl is purified, crystalline cysteine hydrochloride salt (MW 175.6) that is useful as a sulfhydryl standard in assay with Ellman's Reagent, or as an additive in protein refolding experiments.

Thermo Scientific™ Pierce Cysteine-HCl is purified, crystalline cysteine hydrochloride salt (MW 175.6) that is useful as a sulfhydryl standard in assay with Ellman's Reagent, or as an additive in protein refolding experiments.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 25°C.
Product Line Pierce
Form Solid
Quantity 5 g
Product Type Cysteine Hydrochloride
compliance-icons
Product Identifier
  • Pierce Cysteine-HCl
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: L-cysteine

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.