missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Sodium meta-Periodate

Product Code. 11886784 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11886784 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11886784 Supplier Thermo Scientific™ Supplier No. 20504

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gently oxidize glycoprotein carbohydrate sugars to create reactive aldehydes (carbonyls) for crosslinking and immobilization using NaIO4 (CAS 7790-28-5).

Thermo Scientific™ Pierce Sodium meta-Periodate is a gentle oxidizing agent that effectively cleaves cis-diols in carbohydrate sugars to create reactive aldehydes, providing many uses relating to the study and detection of glycoproteins.

Highlights:

  • Capable of the oxidation of cis-diols to aldehydes
  • ACS reagent grade
  • White, crystalline, free-flowing
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store at room temperature.
Label or Dye Aldehyde Group
Label Type Conjugating Groups
Product Line Pierce
Form Powder
Quantity 25 g
Product Type Sodium Meta-Periodate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • SODIUM META-PERIODATE (A.C.S.)
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P370+P378-In case of fire: Use to extinguish.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.