missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Conjugate Purification Kit

Product Code. 10519103 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10519103 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10519103 Supplier Thermo Scientific™ Supplier No. undefined

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gently purify enzyme-conjugated antibodies from unreacted horseradish peroxidase (HRP) or alkaline phosphatase (AP) after labeling.

Thermo Scientific™ Pierce Conjugate Purification Kit is a unique affinity chromatography system for purifying horseradish peroxidase (HRP) or alkaline phosphatase (AP) conjugated antibodies from unreacted enzyme after labeling.

The metal chelate ligand on the column binds the enzyme conjugates while allowing free enzyme to pass through.

Highlights:

  • Reduced background for blotting and ELISA
  • Fewer washes needed to remove background
  • Increased sensitivity of assay system with more active reagent
  • Improved signal:noise ratio of conjugate

Includes:

Immobilized Iminodiacetic acid 1 × 2mL column, Polyacrylamide Desalting Column 1 × 10mL column, FreeZyme Column Activator Buffer 25mL, FreeZyme Elution Buffer 35mL, BupH™ Tris Buffered Saline Pack two each, BupH Phosphate Buffered Saline Pack one each, Activity Preservation Solution 1mL

TRUSTED_SUSTAINABILITY

Specifications

Product Type Conjugate Purification Kit
Quantity 1 Kit
Sufficient For 10 purifications, each of 0.5 to 50 mg of enzyme-conjugated antibody
Content And Storage Sufficient For: 10 purifications, each of 0.5 to 50 mg of enzyme-conjugated antibody
  • IDA Agarose Column, 2 mL, 1 column
  • Polyacrylamide Desalting Column, 10 mL, 1 column
  • Column Activator Buffer, 25 mL
  • Elution Buffer, 35 mL
  • TBS Packs (each makes 500 mL), 2 packs
  • PBS Pack (makes 500 mL), 1 pack
  • Activity Preservation Solution, 1 mL

    Store kit at 4°C.
  • Labeling Scale 0.5 to 3 mg
    Product Line Pierce
    Label or Dye HRP (Horseradish Peroxidase)
    Labeling Target IgG

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.