missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin Coated Plate IP Kit

Product Code. 10425525 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10425525

Brand: Thermo Scientific™ 45360

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Perform immunoprecipitation assays without resins, beads, centrifugation or magnets using coated streptavidin or Protein A/G 96-well microplates.

Thermo Scientific™ Pierce Coated Plate IP Kits enable immunoprecipitation experiments to be performed in 96-well microplates.

These coated microplate immuunoprecipiation kits provide for rapid immunoprecipitation of multiple samples without the usual tedium of pipetting, centrifuging and separating beaded affinity resin in individual microcentrifuge tubes. Immunoprecipitation is accomplished using coated 96-well microplates rather than beaded agarose resin. The plate format allows for faster processing of multiple samples. The Protein A/G Coated Plate Kit can be used for immunoprecipation with any antibody that binds to Protein A or G. The Streptavidin Coated Plate Kit is for immunoprecipitation with biotinylated IP antibodies.

Highlights:

  • Ready-to-use, high quality coated plates provide high capacity and consistency
  • Accommodate antibody and sample volumes comparable to tube-based methods
  • Easily recover immunoprecipitated samples for downstream analysis
  • Coated plates are 96-well strip plates, convenient for experiments requiring only a partial plate
  • Include simple instructions, including detailed explanation of appropriate controls
  • Two kits available, suitable for most common antibody types (mouse, rabbit, human and goat IgG subclasses) or any biotinylated antibody or bait protein

Advantages of the Plate Format:

  • Best suited for simultaneously processing multiple samples and their control conditions
  • Allows faster, easier and more thorough washing than with traditional tube/resin IP methods
  • Makes use of familiar and convenient ELISA tools (multichannel pipettors and plate washing)
  • No no tubes to open and close, and no centrifugation required

Protein A/G Kit (Part No. 45350)

  • Binds all human IgG subclasses; human IgM, IgA (weak), and IgE; and all mouse IgG subclasses
  • Offers increased binding for mouse monoclonal antibodies compared to Protein A or Protein G alone
  • Free of pH dependency when binding IgGs, providing more flexible binding conditions

Streptavidin Kit (Part No. 45360)

  • Used in combination with biotinylated antibodies, resulting in IP and co-IP preparations free of interfering antibody fragments
  • Contains biotin-blocking buffer to minimize endogenous biotin binding

Includes:

Coated 96-well strip plates, uncoated 96-well strip plates, PBS pack, detergent solution, elution buffer, neutralization buffer and sealing tape

Recommended for:

Rapid, trouble-free immunoprecipitation of multiple samples and their controls; Immunoprecipitation when plate-based tools and techniques are preferred

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce Streptavidin Coated Plate IP Kit
Content And Storage Sufficient For: Assaying 48 samples with controls or 192 total IP reactions
• Streptavidin Coated 96-well Strip Plates, 2 plates
• Uncoated 96-well Strip Plate (white), 2 plates
• PBS Pack (makes 500 mL), 1 pack
• Biotin Blocking Buffer, 2 x 15 mL
• Surfact-Amps X-100 Detergent Solution, 6 x 10 mL
• Elution Buffer (pH 2.8), 50 mL
• Neutralization Buffer, 12 mL
• Sealing Tape for 96-well Plates, 18 sheets

Store at 4°C.
Assay Affinity Purification
For Use With (Equipment) Microplate Mixer
Product Line Pierce
Quantity 1 Kit
For Use With (Application) Immunoprecipitation (IP), Pull-Down (Biotin-Based)
Target Not Target-Specific
Type Immunoprecipitation/Co-Immunoprecipitation Kit
Sufficient For 192 samples
Format Kit
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.