missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Co-Immunoprecipitation Kit

Product Code. 10412135 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 reactions kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10412135 50 reactions kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10412135 Supplier Thermo Scientific™ Supplier No. 26149

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Covalently immobilize IP antibodies and perform co-IP assays in which target protein complexes are immunoprecipitated without antibody contamination.

Thermo Scientific™ Pierce Co-Immunoprecipitation Kit provides all the components necessary to perform a properly controlled co-IP experiment and obtain purified products without antibody contamination.

The Co-Immunoprecipitation (Co-IP) Kit enables isolation of native protein complexes from a lysate or other complex mixture by directly immobilizing purified antibodies onto an agarose support. Co-IP is a common approach to study protein:protein interactions that uses an antibody to immunoprecipitate the antigen (bait protein) and co-immunoprecipitate any interacting proteins (prey proteins). Traditional co-IP methods that use Protein A or G result in co-elution of the antibody heavy and light chains that may co-migrate with relevant bands, masking important results. The Pierce Co-IP Kit resolves this issue by covalently coupling antibodies onto an amine-reactive resin. The kit includes optimized buffers for protein binding and recovery, reagents to perform control experiments and efficient spin columns and collection tubes, which shorten the protocol and minimize handling and mixing.

Highlights:

  • Works for any purified antibody – perform co-immunoprecipitation experiments using any purified antibody regardless of species or Ig class (chicken IgY, human IgE, mouse IgG1, IgM, etc.); no dependence on Protein A and Protein G affinity binding

  • No antibody interference – covalent attachment method and non-reducing elution system yields pure co-IP products without contamination by the IP antibody

  • Reusable antibody beads – covalent antibody immobilization allows prepared IP resin to be reused and provides potential savings of costly antibody

  • Complete Kit – all reagents for antibody immobilization and many IP experiments, including binding and wash buffers, elution buffer and convenient microcentrifuge spin columns to make resin manipulations easier and more efficient

  • Control beads included – underivatized agarose beads provided for use as negative control for nonspecific binding

  • Versatile – co-IP method is compatible with any physiolgical (non-denaturing) protein sample buffer that is compatible with the specific antibody-antigen and protein interaction complex

Includes:

AminoLink Plus Coupling Resin, coupling buffer, quenching buffer, wash solution, sodium cyanoborohydride solution, IP lysis/wash buffer, conditioning buffer, Tris-buffered saline, elution buffer, lane marker sample buffer, control agarose resin, spin columns and collection tubes

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: Preparing and performing 50 co-IP reactions, each with 25 μL of resin
• AminoLink Plus Coupling Resin, 2 mL
• Coupling Buffer (20X), 25 mL
• Sodium Cyanoborohydride Solution (5M), 0.5 mL
• Quenching Buffer, 50 mL
• Wash Solution, 60 mL
• IP Lysis/Wash Buffer, 2 x 50 mL
• Modified Dulbecco's PBS (20X), 25 mL
• Conditioning Buffer (100X), 5 mL
• Elution Buffer, 50 mL
• Lane Marker Sample Buffer (5X), 5 mL
• Pierce Spin Columns, Screw Cap, 50 columns
• Microcentrifuge Collection Tubes, 2 mL, 100 tubes
• Microcentrifuge Collection Tubes, 1.5 mL, 50 tubes
• Pierce Control Agarose Resin, 2 mL

Store at 4°C.
Assay Affinity Purification
For Use With (Equipment) Microcentrifuge
Product Line Pierce
Quantity 50 reactions kit
For Use With (Application) Immunoprecipitation (IP)
Target Not Target-Specific
Type Immunoprecipitation/Co-Immunoprecipitation Kit
Sufficient For 50 Reactions
Format Kit
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pierce Co-Immunoprecipitation Kit
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • EUH031-Contact with acids liberates toxic gas.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P273-Avoid release to the environment.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: sodium chloride, sodiumcyanotrihydroborate, sodium hydroxide, trometamol, sodiumchloride, Nonidet P

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.