missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ c-Myc-Tag IP/Co-IP Kit

Product Code. 10571014 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 reactions kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10571014 25 reactions kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10571014 Supplier Thermo Scientific™ Supplier No. 23620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Anti-c-Myc antibody agarose beads and complete reagent set to easily co-immunoprecipitate protein interactions involving c-Myc-tagged fusion proteins.

Thermo Scientific™ Pierce c-Myc Tag IP/Co-IP Kit provides the affinity resin and other reagents necessary to easily perform immunoprecipitation (IP) or co-immunoprecipitation (co-IP) experiments using a c-Myc-tagged protein as the bait.

The c-Myc peptide (EQKLISEEDL) has become a popular fusion tag for mammalian recombinant protein expression, and this kit facilitates immunoprecipitation (IP) or co-immunoprecipitation (co-IP) experiments involving c-Myc-tagged proteins. The kit includes crosslinked beaded agarose to which a highly specific anti-c-Myc antibody is covalently immobilized. Upon incubation with a sample containing the tagged fusion protein, interaction complexes involving the c-Myc-tagged bait protein are captured on the agarose beads. After simple washing steps, the specific protein interaction complex is easily eluted from the resin in the supplied elution buffer or SDS-PAGE sample loading buffer for subsequent analysis. The kit includes the prepared agarose affinity resin, buffers, microcentrifuge spin columns, a positive control and easy-to-follow instructions.

Highlights:

  • Specific – the immobilized anti-c-Myc monoclonal antibody binds the c-Myc epitope tag with high specificity, providing high yield immunoprecipitation products and clean Western blot detection

  • High capacity – excellent results with as little as 1μg of anti-c-Myc antibody in IP mode with the positive control lysate

  • Rigorous – kit includes a positive control lysate that contains over-expressed GST-c-Myc to validate the reagents and specific protocol, and the instructions provide tips for planning other controls

  • Robust – the affinity system is compatible with IP or Co-IP from various cell lysates and physiological (non-denaturing) buffer systems; choose kits with or without mammalian cell lysis buffer

  • Convenient and easy – complete kit includes all necessary reagents, convenient spin columns, and easy-to-follow instructions

Includes:

M-PER™ Mammalian Protein Extraction Reagent (Part No. 23625 only), anti-c-Myc agarose resin, TBS pack, elution buffer, lane marker nonreducing sample buffer, positive control lysate, microcentrifuge spin columns and collection tubes

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce c-Myc-Tag IP/Co-IP Kit
Content And Storage Sufficient For: 25 IP reactions, each using 5 μL resin (5 μg anti-c-Myc antibody)
• Anti-c-Myc Agarose Resin, 250 μL
• c-Myc-tagged Positive Control (Part No. 23633), 500 μL
• TBS Pack (makes 500 mL), 1 pack
• Elution Buffer (pH 2.0), 2 x 25 mL
• Conditioning Buffer (100X), 5 mL
• Non-Reducing Sample Buffer (5X), 5 mL
• Spin Columns and Accessories, 27 columns
• Microcentrifuge Collection Tubes, 100 tubes

Store the c-Myc-tagged Positive Control at -80°C. Store Application Set at 4°C.
Assay Affinity Purification
For Use With (Equipment) Microcentrifuge
Product Line Pierce
Quantity 25 reactions kit
For Use With (Application) Pull-Down (Tag-Based), Immunoprecipitation (IP)
Target Target-Specific
Type Tag IP/Co-IP Kit
Sufficient For 25 Reactions
Format Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.