missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TMTsixplex™ Isobaric Label Reagent Set

Amine-reactive Thermo Scientific TMT Isobaric Mass Tagging Kits and Reagents enable multiplex quantitation of proteins extracted from cells and tissues from up to 6 samples using tandem mass spectrometry.

Brand:  Thermo Scientific™ 90066

Code : PN

Additional Details : Weight : 2.72160kg

 View more versions of this product

Product Code. 10322094

  • £1992.00 / Each
Estimated Shipment: 07-05-2024
to see stock.
Add to basket

Description

Description

Tandem Mass Tag™ 6-plex (TMTsixplex™) Reagents are sets of isobaric compounds (i.e., same mass and structure, also called isotopomers) that are NHS-activated for covalent, irreversible labeling of primary amines (–NH2) groups. Each isobaric reagent contains a different number of heavy isotopes in the mass reporter region, which results in a unique reporter mass during tandem MS/MS for sample identification and relative quantitation. Amine-reactive Thermo Scientific TMT Isobaric Mass Tagging Kits and Reagents enable multiplex quantitation of proteins extracted from cells and tissues from up to 6 samples using tandem mass spectrometry.

Features of TMTsixplex Label Reagents and Kits:
Powerful—multiplexed MS analysis increases sample throughput and enables relative quantitation of up to six different samples derived from cells, tissues or biological fluids
Robust—consistent chemistry allows efficient transition from method development to multiplex quantitation, enabling biomarker discovery research
Efficient—amine-reactive NHS-ester activated reagents ensures greater than 95% labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
Compatible—optimized for use with high resolution Thermo Scientific Orbitrap MS/MS platforms, with data analysis fully supported by Proteome Discoverer™ 2.3 and above

Applications of TMTsixplex Label Reagents and Kits:
• Protein identification and quantitation from multiple samples of cells, tissue or biological fluids
• Protein expression profiling of normal vs. disease states or control vs. treated
• Quantitative analysis of proteins for which no antibodies are available
• Identification and quantitation of membrane and post-translationally modified proteins
• Identification and quantitation of hundreds to thousands of proteins in a single experiment

Since TMT reagents share an identical structure and mass (i.e., isotopomers), labeled peptides co-elute during LC separation and are co-isolated during MS/MS analysis, resulting in fewer missing peptide identifications among samples. During MS/MS analysis, each isobaric tag is also fragmented to produce a unique reporter ion mass that is used for sample identification and quantitation. Protein quantitation is accomplished by comparing the relative intensities of the six reporter ions.

TMT Reagents are provided as standalone sets or in optimized kit formats containing all necessary reagents and controls for maximum flexibility, convenience, and reliability. Tandem Mass Tag Reagents combined with high resolution Thermo Scientific Orbitrap instruments and Proteome Discoverer software provide integrated total system solutions for quantitative protein expression analysis. For high resolution analysis of the TMT-labeled peptides, the recommended LC column for Nanospray Flex source is Acclaim PepMap 100 C18 LC Column (Cat. No. 164942 or 164939). For EASY-Spray source, the recommended LC column is EASY-Spray C18 LC Column (Cat. No. ES902 or ES903).

Specifications

Specifications

TMTsixplex™ Isobaric Label Reagent Set, 5 x 0.8 mg
TMTsixplex™
Tandem Mass Tag™ (TMT) Labels
Protein
Protein Labeling
Mass Spectrometer
Sufficient for: Five sixplex (5 x 6-way) experiments
• TMT6-126 Label Reagent, 5 x 0.8 mg
• TMT6-127 Label Reagent, 5 x 0.8 mg
• TMT6-128 Label Reagent, 5 x 0.8 mg
• TMT6-129 Label Reagent, 5 x 0.8 mg
• TMT6-130 Label Reagent, 5 x 0.8 mg
• TMT6-131 Label Reagent, 5 x 0.8 mg

Store at –20°C.

Cell Lysate
Mass Spectrometry
5 x 0.8 mg (per tag)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.