missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ AMAS (N-α-maleimidoacet-oxysuccinimide ester)
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce AMAS is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a very short spacer arm (4.4 Ås).
Highlights:
- Reactive groups: NHS ester and maleimide
- Reactive toward: amino and sulfhydryl groups
Specifications
Specifications
| Chemical Reactivity | Amine-Sulfhydryl |
| Cleavable | No |
| Description | AMAS |
| Molecular Weight (g/mol) | 252.18 |
| PEGylated | No |
| Spacer Arm Length | 4.4 Å |
| Content And Storage | Upon receipt store desiccated at 4°C. |
| Cell Permeability | Yes |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?