missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Proteinase K Solution, ChIP grade

Product Code. 10504885 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.25 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10504885 0.25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10504885 Supplier Thermo Scientific™ Supplier No. 26160

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chromatin immunoprecipitation (chromatin IP, ChIP) assay kit containing all reagents, agarose affinity beads, and a simple, fast, reliable protocol.

Thermo Scientific™ Pierce Agarose ChIP Kit provides a complete set of reagents and a simple, fast and reproducible protocol to perform chromatin immunoprecipitation (ChIP) assays.

This chromatin IP kit contains sufficient reagents to perform 30 ChIP assays with appropriate controls using an optimized protocol and a convenient spin-column format. The kit includes a complete Chromatin Prep Module (also available separately) to efficiently extract and solubilize crosslinked DNA-protein interaction complexes from cells, plus an immunoprecipitation module that contains high-capacity, ChIP-grade Protein A/G agarose.

Highlights:

  • Simple and streamlined ChIP assay protocol (< 8 hours)
  • Highly efficient isolation and lysis of nuclei
  • Easy and reproducible enzymatic digestion
  • Low-background and high-capacity Protein A/G agarose resin
  • Highly specific positive control reagents included: RNA polymerase II antibody and GAPDH PCR primers
  • Fast and reproducible spin-column format; columns and collection tubes included
  • High-recovery DNA purification
  • Choose a complete ChIP assay kit or the separate chromatin preparation module

Includes:

Kit contains the Chromatin Prep Module (Part No. 26158; 11 components) for chromatin extraction and a complete set of immunoprecipitation reagents (15 components) for IP experiments with controls

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce Proteinase K
Content And Storage Upon receipt store at -20°C.
Form Liquid
Quantity 0.25 mL
Type Proteinase K (ChIP grade)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.