missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PID1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | PID1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunoprecipitation 1:500 - 1:1000 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
18633530
|
Novus Biologicals
NBP2-93820-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679220
|
Novus Biologicals
NBP2-93820-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PID1 Polyclonal antibody specifically detects PID1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpécification
| PID1 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Endocrinology, metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 55022 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunoprecipitation 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| HMFN2073, P-CLI1, phosphotyrosine interaction domain containing 1, PTB-containing, cubilin and LRP1-interacting protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 124-248 of human PID1 (NP_060403.3). REDVFPANALLEIRPFQVWLHHLDHKGEATVHMDTFQVARIAYCTADHNVSPNIFAWVYREINDDLSYQMDCHAVECESKLEAKKLAHAMMEAFRKTFHSMKSDGRIHSNSSSEEVSQELESDDG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit