missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PID1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93820-0.02ml
This item is not returnable.
View return policy
Description
PID1 Polyclonal antibody specifically detects PID1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| PID1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunoprecipitation 1:500 - 1:1000 | |
| HMFN2073, P-CLI1, phosphotyrosine interaction domain containing 1, PTB-containing, cubilin and LRP1-interacting protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 124-248 of human PID1 (NP_060403.3). REDVFPANALLEIRPFQVWLHHLDHKGEATVHMDTFQVARIAYCTADHNVSPNIFAWVYREINDDLSYQMDCHAVECESKLEAKKLAHAMMEAFRKTFHSMKSDGRIHSNSSSEEVSQELESDDG | |
| 0.02 mL | |
| Cell Biology, Cell Cycle and Replication, Endocrinology, metabolism | |
| 55022 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction