missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIB5PA Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94054-0.02ml
This item is not returnable.
View return policy
Description
PIB5PA Polyclonal antibody specifically detects PIB5PA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PIB5PA | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100 | |
| A, inositol polyphosphate 5-phosphatase J, inositol polyphosphate-5-phosphatase J, phosphatidylinositol 4,5-bisphosphate 5-phosphatase A, PIPP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-360 of human INPP5J (NP_001002837.1). VTWNVGTAMPPDDVTSLLHLGGGDDSDGADMIAIGLQEVNSMLNKRLKDALFTDQWSELFMDALGPFNFVLVSSVRMQGVILLLFAKYYHLPFLRDVQTDCTRTGLGGYWGNKGGVSVRLAAFGHMLCFLNCHLPAHMDKAEQRKDNFQTILSLQQFQGPGAQGILDHDLVFWFGDLNFRIESYDLHFVKFAIDSDQLHQLWEKDQLNMAKNTWPILKGFQEGPLNFAPTFKFDVGTNKYDTSAKKRKPAWTDRILWKVKAPGGGPSPSGRKSHRLQVTQHSYRSHMEYTVSDHKPVAAQF | |
| 0.02 mL | |
| Lipid and Metabolism, Protein Phosphatase | |
| 27124 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction