missing translation for 'onlineSavingsMsg'
Learn More

PIB5PA Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£149.00 - £366.00

Specifications

Antigen PIB5PA
Dilution Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18696722
View Documents
Novus Biologicals
NBP2-94054-0.02ml
0.02 mL
£149.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18643412
View Documents
Novus Biologicals
NBP2-94054-0.1ml
0.1 mL
£366.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PIB5PA Polyclonal antibody specifically detects PIB5PA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

PIB5PA
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Lipid and Metabolism, Protein Phosphatase
PBS (pH 7.3), 50% glycerol
27124
IgG
Affinity purified
Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100
Polyclonal
Purified
RUO
Human, Mouse, Rat
A, inositol polyphosphate 5-phosphatase J, inositol polyphosphate-5-phosphatase J, phosphatidylinositol 4,5-bisphosphate 5-phosphatase A, PIPP
Recombinant fusion protein containing a sequence corresponding to amino acids 60-360 of human INPP5J (NP_001002837.1). VTWNVGTAMPPDDVTSLLHLGGGDDSDGADMIAIGLQEVNSMLNKRLKDALFTDQWSELFMDALGPFNFVLVSSVRMQGVILLLFAKYYHLPFLRDVQTDCTRTGLGGYWGNKGGVSVRLAAFGHMLCFLNCHLPAHMDKAEQRKDNFQTILSLQQFQGPGAQGILDHDLVFWFGDLNFRIESYDLHFVKFAIDSDQLHQLWEKDQLNMAKNTWPILKGFQEGPLNFAPTFKFDVGTNKYDTSAKKRKPAWTDRILWKVKAPGGGPSPSGRKSHRLQVTQHSYRSHMEYTVSDHKPVAAQF
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.