missing translation for 'onlineSavingsMsg'
Learn More

Ovalbumin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications

£205.00 - £577.00

Specifications

Antigen Ovalbumin
Classification Polyclonal
Conjugate Unconjugated
Form Antisera
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18770984
View Documents
Novus Biologicals
NB600-922SS
0.02 mg
£205.00
0.02mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18077465
View Documents
Novus Biologicals
NB600-922-0.1MG
0.1 mg
£577.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ovalbumin Polyclonal specifically detects Ovalbumin in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Single Cell Western.
Specifications

Specifications

Ovalbumin
Unconjugated
Rabbit
Chicken, Virus
Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8
IgG
Polyclonal
Antisera
RUO
P01012
396058
Primary
Videoer
SDS
Dokumenter

Dokumenter

Produktcertifikater

For Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.