missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ovalbumin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB600-922-0.1MG
This item is not returnable.
View return policy
Description
Ovalbumin Polyclonal specifically detects Ovalbumin in Chicken, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Single Cell Western.
Specifications
| Ovalbumin | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8 | |
| Rabbit | |
| Ion exchange chromatography | |
| RUO | |
| 396058 | |
| Chicken, Virus | |
| Antisera |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:5000 - 1:20000, Immunohistochemistry 1:300 - 1:2000, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:300 - 1:2000 | |
| P01012 | |
| SERPINB14 | |
| Ovalbumin [hen egg white] | |
| 0.1 mg | |
| Primary | |
| Reacts with ovalbumin. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction